Documenti fondamentali
Z222372
Metal standard taper clip
joint size, 19
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.35
Descrizione generale
Nickel-plated spring-steel joint clips offer increased strength and resistance to heat and cracking over Delrin® plastic type. Securely holds conical joints under pressure at temperatures up to 392 °F (200 °C). Will not scratch glassware.
Note legali
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)