Z207187
Broken-glass disposal boxes
Benchtop
Sinonimo/i:
benchtop waste glass box, laboratory waste glass box
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.35
Prodotti consigliati
Materiali
cardboard
Caratteristiche
Holds broken glass
Confezionamento
pkg of 6 ea
Produttore/marchio commerciale
Bel-Art F24653-0002
Disponibilità
not available in EU
Largh. × lungh. × alt.
8 in. × 8 in. × 10 in.
Descrizione generale
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full, safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English, French, Spanish, and German). Shipped flat.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)