Z202746
Jacketed beaker
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
Caratteristiche
graduations: no
Volume
100 mL
Categorie correlate
Descrizione generale
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.
Outer Jacket O.D. 57mm x H 120mm.
Inner Vessel I.D. ~38mm x D 110mm.
Inner Vessel I.D. ~38mm x D 110mm.
Altre note
Browse all glass beakers
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z202746-1EA | 4061837617379 |