Documenti fondamentali
Z137073
Jacketed beaker
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Materiali
borosilicate glass
Caratteristiche
graduations: no
Volume
400 mL
Descrizione generale
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.
Altre note
Browse all glass beakers
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z137073-1EA4061832347301