Z163961
Gravity column
Short
Sinonimo/i:
Gravity Filter
About This Item
Codice UNSPSC:
23151816
NACRES:
NB.51
Prodotti consigliati
Giunto
joint ST/NS24/40
Lungh. × D.I. × D.E.
33 cm × 24 mm × 28 mm
Categorie correlate
Descrizione generale
Gravity column can prevent special moment frame structures from collapsing.
Caratteristiche e vantaggi
- Provided with a 4mm polytetrafluoroethylene (PTFE) stopcock
- Has no frit and uses a glass-wool plug and sand
- Use along with coarse silica gel (e.g., Cat. No. 60744) for fast and general-purpose purification.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)