Documenti fondamentali
Z135704
Sigma-Aldrich
Scienceware® Polystop dropper bottle
capacity 125 mL
Sinonimo/i:
bottle dropper, dropper bottles
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
colorless bottle
glass bottle
round bottle
Giunto
joint ST/NS neck
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F11604-0125
Capacità
125 mL
Descrizione generale
Use dropper without removing stopper. Clear glass bottle. Dropper with PP stopper and soft-vinyl bulb. Ground, tapered neck on bottle fits stopper. Can be disassembled to clean parts separately.
Note legali
Polystop is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)