Documenti fondamentali
Y906522
Sigma-Aldrich
Valve
High Pressure, stainless steel diaphragm, 1/4 in. NPT straight path, external
About This Item
Codice UNSPSC:
12142200
NACRES:
NA.12
Materiali
stainless steel diaphragm
Livello qualitativo
Parametri
3000 psi max. pressure
Lunghezza
2.387 in.
Peso
214 g
Raccordi
1/4 in. NPT straight path, external
Categorie correlate
Altre note
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)