Documenti fondamentali
Y906468
Sigma-Aldrich
Valve
stainless steel diaphragm, for CGA 660 right angle path, external, right hand thread
About This Item
Codice UNSPSC:
12142200
NACRES:
NA.12
Materiali
stainless steel diaphragm
Livello qualitativo
Parametri
2640 psi max. pressure
Lunghezza
4.62 in.
Peso
560 g
Raccordi
for CGA 660 right angle path, external, right hand thread
Categorie correlate
Altre note
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)