Documenti fondamentali
WHAWB100040
Unicore Punch Kit
I.D. 6.0 mm, pkg of 4 ea
Sinonimo/i:
UniCore Punch Kit 6.00 mm (4), Whatman FTA, Z747432, cutting mat, harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
Codice UNSPSC:
41105300
NACRES:
NB.22
Confezionamento
pkg of 4 ea
Produttore/marchio commerciale
Qiagen WB100040
Diam. int.
6.0 mm
Descrizione generale
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Manual UniCore punch system for sample disc removal from FTA cards.
A cutting mat (also available separately: WHAWB100022) included in UniCore Punch Kits ensures clean sample cuts from FTA/FTA Elute Cards and extends the life time of the cutting tip (up to 500 punches).
Caratteristiche e vantaggi
  • Stainless steel cutting edges Stainless steel cutting edges (6.0 mm) enable manual coring, retrieval, and controlled ejection of the FTA punch.
  • Designed to minimize contamination and carryover between samples when a cleaning procedure is followed.
Altre note
Field of Use : For internal research use only
Note legali
FTA is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.