Documenti fondamentali
WHA1703050
Whatman® ashless filter aids
Sinonimo/i:
Whatman filter, Z740764, filter papers
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Sterilità
non-sterile
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Whatman 1703-050
Whatman Article No. 28418284 (US reference)
Descrizione generale
Whatman ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive ‘prefilter′ layer on top of normal filter paper.
Whatman ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive “prefilter” layer on top of normal filter paper.
Easily dispersible, the powder is of a purity similar to that of Whatman ashless quantitative papers. It is supplied with a two-ended scoop for measuring 0.5 g or 2.5 g quantities.
Caratteristiche e vantaggi
  • Easily dispersible, the clippings are of purity similar to that of Whatman ashless quantitative papers.
  • Supplied with a two-ended scoop for measuring 0.5 g or 2.5
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)