WHA10498761
Whatman® multiple vacuum filtration device
3-place, 3-place manifold, stainless steel
Sinonimo/i:
Z752177
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Prodotti consigliati
Materiali
stainless steel
Confezionamento
pk of 1
Produttore/marchio commerciale
Whatman 10498761
Whatman Article No. 28416533 (US reference)
Categorie correlate
Descrizione generale
Stainless steel filter funnel manifold, for microbiology monitors and analytical funnels.
Caratteristiche e vantaggi
- The stainless steel manifold for three or six filtration units is fitted with stainless steel units.
- The apparatus can be autoclaved and sterilized by dry heat at up to 180°C.
- Suitable only for vacuum operation.
- If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)