WHA10310992
Whatman® drying pad
Sinonimo/i:
Z695513
About This Item
Codice UNSPSC:
40161507
NACRES:
NB.24
Prodotti consigliati
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Whatman 10310992
Whatman Article No. 28414185 (US reference)
Largh. × lungh.
37 mm × 100 mm
Descrizione generale
Absorbent paper strips for the removal of excess liquids, e.g., from microscope specimens.
Caratteristiche e vantaggi
- Highly absorbent
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)