Documenti fondamentali
THSD1146
ThalesNano MidiCart catalyst cartridge system
L × diam. 116 mm × 9.2 mm, Raney cobalt
Sinonimo/i:
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Codice UNSPSC:
41103817
NACRES:
NB.51
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
ThalesNano THS D1146
L × diam.
116 mm × 9.2 mm
Descrizione generale
MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.
Note legali
MidiCart is a trademark of ThalesNano Nanotechnology Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)