Documenti fondamentali
SYNV210250
Synthware Dewar type vacuum trap
250 mL, stopcock: PTFE
Sinonimo/i:
Synthware V210250
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware V210250
Capacità
250 mL
Descrizione generale
Dry ice, Dewar style trap condenser designed for large volume solvent removal. Choice either glass stopcock for better vacuum or PTFE stopcock for grease-free operation. With 10 mm hose connections.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)