Documenti fondamentali
SYNV165050
Synthware vacuum trap with serrated hose connections
joint: ST/NS 50/50, body O.D. 55 mm, L 275 mm , below joint
Sinonimo/i:
Synthware V165050
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Giunto
joint: ST/NS50/50
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware V165050
Lunghezza
275 mm , below joint
Diam. est. corpo
55 mm
Descrizione generale
With joints for easy removal of condensate. Serrated hose connections for side tube have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration. Center tube hose connection is always 10mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)