Documenti fondamentali
SYNV131005
Synthware conical bottom reaction vial with holed compression cap
5 mL, joint: ST/NS 14/10
Sinonimo/i:
Synthware V131005, reaction tube
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS14/10
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware V131005
Capacità
5 mL
Categorie correlate
Descrizione generale
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)