Documenti fondamentali
SYNM401004
Synthware all-glass vacuum/inert gas manifold with hollow high vacuum stopcocks
number of ports, 4, Hose Connections: Front-right, Rear-left
Sinonimo/i:
Synthware M401004, vacuum manifold, vacuum unit
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Caratteristiche
number of ports 4
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M401004
Descrizione generale
With 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mmO.D. with front-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)