SYNA751420
Synthware™ 90 degree vacuum adapter with glass stopcock
male joint: ST/NS 14/20
Sinonimo/i:
Synthware A751420, flow control adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
male joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A751420
Categorie correlate
Descrizione generale
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNA751420-1EA | 4061832207537 |