SYNF611960
Synthware™ pressure equalizing addition funnel with metering stopcock
60 mL, joint: ST/NS 19/22
Sinonimo/i:
Synthware F611960, dropping funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F611960
Capacità
60 mL
60 mL
Categorie correlate
Descrizione generale
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)