SYNF381420M
Synthware™ filter funnel with fritted disc
joint: ST/NS 14/20, disc porosity: medium
Sinonimo/i:
Synthware F381420M
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.51
Prodotti consigliati
Materiali
borosilicate glass
Giunto
joint: ST/NS14/20
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
F381420M
Diam. disco
20 mm
Diam. filtro
20 mm
Categorie correlate
Descrizione generale
Synthware™ filter funnel features a vacuum port and medium filter disc (O.D. 20 mm) porosity designed for efficient filtration. The vacuum port facilitates rapid filtration by creating a vacuum, enhancing the separation process for various applications. With upper standard taper outer joint (14/20) to fit various laboratory glassware.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)