SYNF321025
Synthware™ pear shaped flask
25 mL, joint: ST/NS 14/10
Sinonimo/i:
Synthware F321025, pear shape glass flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Prodotti consigliati
Materiali
borosilicate glass flask
pear-shaped
single-neck
Giunto
joint: ST/NS14/10
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F321025
Capacità
25 mL
25 mL
Categorie correlate
Descrizione generale
Supplied with one compression cap, one Viton® O-ring and one PTFE –faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNF321025-1EA | 4061832186849 |