Documenti fondamentali
SYNF291210
Synthware solvent addition funnel
top diam. 100 mm
Sinonimo/i:
Synthware F291210, glass funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F291210
Diametro sommità
100 mm
Categorie correlate
Descrizione generale
Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica-gel/packing layer. Lower end of the funnel is closed and the stem has a series of small holes through which the solvent drains. Top of the funnel is 90mm in diameter with the stem size as listed.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)