Documenti fondamentali
SYNF10401LA
Synthware 47 mm filtration apparatus with stainless steel support, complete
joint: ST/NS 40/35
Sinonimo/i:
Synthware F10401LA, complete filtration apparatus
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Giunto
joint: ST/NS40/35
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F10401LA
Categorie correlate
Descrizione generale
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has removable 47 mm stainless steel support screen for use with viscous or proteinaceous solutions. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)