Documenti fondamentali
SYNC164250
Synthware vacuum jacketed distillation column with indents to support packing material
joint: ST/NS 24/40, jacket H 250 mm
Sinonimo/i:
Synthware 164250, glass distilling column
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C164250
Altezza camicia
250 mm
Descrizione generale
With 12 mm I.D. indentations to support packing material. Vacuum jacketed to improve performance .
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)