Documenti fondamentali
SYNA201924
Synthware reducing adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 24/40
Sinonimo/i:
Synthware A201924, glass reducing adapter, connecting adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
bottom joint: ST/NS24/40
joint: ST/NS top19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A201924
Descrizione generale
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (19/22).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)