Documenti fondamentali
SYNA202434
Synthware reducing adapter
top joint: ST/NS 24/40, bottom joint: ST/NS 34/45
Sinonimo/i:
Synthware A202434, glass reducing adapter, connecting adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
bottom joint: ST/NS34/45
joint: ST/NS top24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A202434
Descrizione generale
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (34/45) is larger than the top joint (24/40).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)