SLW1000/30D
Pyrex® Griffin beakers, low form, with printed trace code
3000 mL
Sinonimo/i:
Pyrex, SciLabware 1000/30D
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
white graduation
Caratteristiche
low form
with printed trace code
Produttore/marchio commerciale
1000/30D
SciLabware
Alt. × Diam.
210 mm × 150 mm
Capacità
3000 mL
Descrizione generale
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex borosilicate glass
- Excellent thermal performance
- High resistance to chemical attack
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- White graduations and marking spot
Caratteristiche e vantaggi
- Complies with ISO 3819 and DIN 12331
- Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
- Excellent thermal performance
- High resistance to chemical attack
- Printed with Trace Code for downloadable batch certificate
- White graduations and marking spot
- Graduations are approximate
- 1000/38D is supplementary to ISO range
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)