MT51343192
Mettler Toledo InLab® Reference Flow electrode
Reference electrode with triple ceramic junction
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Prodotti consigliati
Materiali
glass shaft
Descrizione
S7 connector
Without cable
Produttore/marchio commerciale
Mettler-Toledo 51343192
Campo di misura della temperatura
0-130 °C
Lungh. × largh.
120 mm × 12 mm
Descrizione generale
Mettler Toledo InLab® Reference Flow is a glass-body, reference electrode with S7 screw head. Filled with 3 mol/L KCl for routine applications
- Triple ceramic diaphragm
- Electrolyte refill hose coupling
Note legali
InLab is a registered trademark of Mettler-Toledo, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)