Documenti fondamentali
MT51341000
Mettler Toledo DX series GSE half-cells electrode
GSE Ammonia
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Materiali
shaft (PPS)
Produttore/marchio commerciale
Mettler-Toledo 51341000
Campo di misura
0.5 ... 5.0-3
Campo di misura della temperatura
0-50 °C
pH
>11
Descrizione generale
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your GSE half-cell, and you have a new GSE!
Some interference may be seen with volatile bases. ISA solution: 1 mol/L KNO3
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)