Documenti fondamentali
MT30027776
Mettler Toledo electrode cable
connector: K8SD, Mini-LTW, L 1.2 m
Sinonimo/i:
Mettler-Toledo electrode cable
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Produttore/marchio commerciale
Mettler-Toledo 30027776
Lunghezza
1.2 m
Descrizione generale
Dedicated cable to connect InLab® Smart Pro-ISM to SevenCompact S220, K8SD to Mini-LTW, 1.2 m
Mettler Toledo electrodes can easily be connected to various third-party instruments. All you have to do is select the appropriate cable. We offer a wide choice of cable lengths and plug combinations. For Mettler Toledo reference electrodes and temperature probes.
Note legali
InLab is a registered trademark of Mettler-Toledo, Inc.
Prodotti correlati
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)