Documenti fondamentali
DWKWCL1000AD-3
WHEATON® CELLine flasks for adherent cell culture
polystyrene, flask capacity (1,000 mL), case of 3 ea
Sinonimo/i:
Adherent Cell Culture Flasks, CELLine Flasks, Cell Culture Flasks
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.13
Materiali
molded polypropylene neck
polystyrene
screw top plastic cap
Sterilità
sterile; γ-irradiated
Caratteristiche
with cap
Confezionamento
case of 3 ea
Produttore/marchio commerciale
WHEATON® WCL1000AD-3
Capacità pallone
(1,000 mL)
Descrizione generale
CELLine AD Bioreactor Flask for high density adherent cell culture.
Media compartment with cell growth compartment for antibody and protein production.
Use with Hybridoma, Insect, CHO, NSO, HEK, BHK cell lines
  • Disposable and ready-to-use
  • High call density and high product concentration
  • Bulk media chamber allows for longer run times without media refreshing and reduces operation time
  • Decreases use of consumables
  • Cost-efficient, space saving, and stackable
  • No additional equipment required for operation
Caratteristiche e vantaggi
  • Ideal for high density cell culture, designed to enhance small scale bio-production for antibody and protein generation_x000D_
  • Available in 1000ml flask for suspension (CELLine 1000) or adherent (CELLine 1000-AD) cell culture and 350ml flask for suspension culture only_x000D_
  • Disposable and ready-to-use_x000D_
  • Multi-harvest operation ensures a cost efficient solution for antibody and protein production_x000D_
  • Ideal for use with cell lines such as Hybridoma, CHO, HEK, BHK, Sf9
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)