Documenti fondamentali
DWKW217922
Wheaton wide-mouth bottle
clear soda-lime glass bottle, capacity (1,000 mL), white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Sinonimo/i:
W217922
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Diametro bottiglia × alt.:
(98 mm) (174 mm)
Capacità:
(1,000 mL)
Materiali:
(PTFE faced LDPE liner)
clear soda-lime glass bottle
white polypropylene cap
Caratteristiche:
with cap
with cap
Raccordi:
89-400
Confezionamento:
case of 12 ea
Materiali
(PTFE faced LDPE liner)
clear soda-lime glass bottle
white polypropylene cap
Caratteristiche
with cap
with cap
Confezionamento
case of 12 ea
Produttore/marchio commerciale
WHEATON® W217922
Diametro bottiglia × alt.
(98 mm) (174 mm)
Capacità
(1,000 mL)
Raccordi
89-400
Descrizione generale
The Wheaton wide-mouth bottle is the largest size available in Type III glass bottles, specifically designed for general sample storage. Its wide-mouth opening facilitates easy filling and pouring, making it convenient for handling various types of samples. Constructed from durable glass, this bottle provides excellent chemical resistance, ensuring the integrity of the stored contents. Ideal for laboratories and industrial applications, it offers a reliable solution for managing bulk samples and liquids efficiently.
Caratteristiche e vantaggi
• Available in larger bottle sizes• Wide mouth / neck• Clear, Type III soda-lime glass• Available with caps attached or without caps• HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification• Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)