DWK92812G-4000
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with glass stopper
glass flask, flask capacity (4,000 mL), class A
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
glass flask
glass stopper
Caratteristiche
class A
Confezionamento
case of 1 ea
Produttore/marchio commerciale
Kimble® 92812G-4000
Capacità pallone
(4,000 mL)
Descrizione generale
Class A Heavy Duty Wide Mouth Volumetric Flasks with Glass Stopper
- Pipet access is easy with wide-mouth volumetric flasks
- Superior strength, durability and safety from heavy, uniform walls
- Quick identification with large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Standard Taper glass stoppers
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Caratteristiche e vantaggi
- Manufactured from borosilicate glass 3.3
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Heavy duty walls provide superior strength, durability and safety
- Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
- Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
- Printed large volume numbers allow quick identification when selecting glassware
- Flat interior bottoms for stirring with standard stir bars
- Calibrate to contain
- Graduated to Class A volumetric tolerances
- Supplied with or without stopper
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)