DWK227494-02
Wheaton® Bod Bottle
capacity 60 mL, bottle diam. × H 43 mm × 115 mm, glass cap (Robotic Stopper)
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
glass cap (Robotic Stopper)
Confezionamento
pkg of 36 ea
Produttore/marchio commerciale
WHEATON® 227494-02
Diametro bottiglia × alt.
43 mm × 115 mm
Capacità
60 mL
Descrizione generale
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Caratteristiche e vantaggi
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
DWK227494-02-36EA |