Documenti fondamentali
DWK11-5200-300
Wheaton® Microliter Vials With Inserts Installed
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
Materiali
clear vial
Caratteristiche
vial type with Marking Patch with 11-0000-300 inserts
Confezionamento
pkg of 1 packs
Produttore/marchio commerciale
WHEATON® 11-5200-300
Applicazioni
Chromatography
Caratteristiche e vantaggi
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)