Documenti fondamentali
CLS7095BNMR
Corning® Pasteur pipettes, non-sterile
L 13 1/4 in. (337 mm), long tip, soda lime
Sinonimo/i:
Pipettes, pipettes, serological pipets, serological pipettes
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.04
Materiali
borosilicate glass
Sterilità
non-sterile
Caratteristiche
graduations: no
subdivision: no
Confezionamento
case of 100 ea
pack of 100 ea
Produttore/marchio commerciale
Corning 7095B-NMR
Lunghezza
13 1/4 in. (337 mm)
Descrizione generale
  • Corning® disposable Pasteur pipettes are made of sturdy soda-lime glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.
Disposable. Features a constriction one inch below the top to facilitate plugging. Ideal for rapid nonvolumetric work in bacteriology, immunology, hematology, serology studies, blood bank and clinical chemistry procedures.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Pipette Pasteur Sigma non-plugged, L 5 3/4 in.
S6018
Pipette Pasteur Sigma
Pipette Pasteur Sigma cotton plugged, L 9 in.
S6143
Pipette Pasteur Sigma
BRAND® dropping pipettes capacity 5 mL, with integrated bellows
Z331805
BRAND® dropping pipettes
BRAND® pipette withdraw volume 5.5 mL (including bulb), graduations, 0.25 mL
BR747775
BRAND® pipette
Volac® Pasteur pipette Cotton plugged, L 230 mm
Z310751
Volac® Pasteur pipette