CLS70582X
Pyrex® disposable graduated bacteriological pipette, sterile
volume 2.2 mL
Sinonimo/i:
pipet
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.43
Prodotti consigliati
Materiali
Pyrex
Descrizione
plugged
Produttore/marchio commerciale
Corning 7058-2X
Campo di misura del volume
2.2 mL
Capacità
2.2 mL
Categorie correlate
Descrizione generale
This pipet is very convenient when duplicate platings are to be made on the same medium or for plating differential medium for the determination of gram-negative bacteria, mold and yeast, or total plate count. Both sizes are designed for gravity feed (blow-out is not required).
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)