Documenti fondamentali
CLS3160102
Pyrex® petri dishes
complete, O.D. × H 100 mm × 20 mm
Sinonimo/i:
Corning® 3160
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.43
Materiali
borosilicate glass
Sterilità
non-sterile
Confezionamento
pack of 12
Produttore/marchio commerciale
Corning 3160-102
D.E. × alt.
100 mm × 20 mm
Descrizione generale
  • PYREX® culture dishes are made of clear borosilicate glass.
  • They can withstand repeated sterilization (wet or dry) and are chemically resistant.
  • Beaded edges provide greater mechanical strength and reduce capillary action of condensed moisture.
  • Top marked in blue enamel, bottom in white, enables easier sorting.
  • Bottoms include a triangular, enamel reference point for serial dilutions.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)