CLS17304H
Corning® Snap-Seal plastic sample containers
volume 120 mL
Sinonimo/i:
storage vessel
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.11
Prodotti consigliati
Materiali
translucent polypropylene
Produttore/marchio commerciale
Corning 1730-4H
diam. × alt.
45 mm × 91 mm
Volume
120 mL
Descrizione generale
Snap-Seal locking device keeps cap closed and secure, for reliable leak-tight seal when properly closed.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)