CAD6056
3-way stopcock
female Luer lock to female Luer lock, male Luer lock, in. Y in. type
About This Item
Codice UNSPSC:
41122000
NACRES:
NB.03
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Cadence Science 6056
Categorie correlate
Descrizione generale
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
- Made from nickel plated brass or stainless steel
- Tested to 45 psi water pressure
- High pressure stopcocks are rated to 290 psi
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6056-1EA | 4061832517292 |