BR92739
BRAND® Erlenmeyer flask with beaded rim and graduation, narrow mouth
volume 300 mL
Sinonimo/i:
Erlenmeyer flasks, Flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
flat bottom
Giunto
joint neck34 mm
Sterilità
non-sterile
Caratteristiche
without cap
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
BRAND 92739
Capacità
300 mL
Volume
300 mL
Descrizione generale
The BRAND® Erlenmeyer Flask with beaded rim and graduations is a high-quality laboratory vessel designed for versatile use in chemical and biological applications. Made from Boro 3.3 glass, this flask meets DIN ISO 1773 standards, ensuring durability and reliability in various laboratory environments.
Caratteristiche e vantaggi
- Good mechanical resistance
- Easy to clean
- Diverse applications
Altre note
Browse all Erlenmeyer flasks
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)