Documenti fondamentali
BR92717
BRAND® Erlenmeyer flask with beaded rim and graduation, narrow mouth
volume 50 mL
Sinonimo/i:
Erlenmeyer flasks, Flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flat bottom
Giunto
joint neck22 mm
Sterilità
non-sterile
Caratteristiche
without cap
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
BRAND 92717
Capacità
50 mL
Volume
50 mL
Descrizione generale
The BRAND® Erlenmeyer Flask with beaded rim and graduations is a high-quality laboratory vessel designed for versatile use in chemical and biological applications. Made from Boro 3.3 glass, this flask meets DIN ISO 1773 standards, ensuring durability and reliability in various laboratory environments.
Caratteristiche e vantaggi
  • Good mechanical resistance
  • Easy to clean
  • Diverse applications
Altre note
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
D210404-25G4061833561584
D210404-500G4061833561591
D210404-5G
D4630-10G
D4630-1KG4061833565568
D4630-50G
D4630-100G4061833565551
D4630-25G
D4630-500G4061833553473
D4630-5G
D210404-100G4061833561577
BR92717-10EA4061831861716