BR87616
BRAND® PP beaker with spout, low form
volume 500 mL
Sinonimo/i:
Beakers, Plastics
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Descrizione
ISO 7056
Caratteristiche
graduations
beaded rim and spout
Produttore/marchio commerciale
BRAND 87616
Volume
500 mL
Categorie correlate
Descrizione generale
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Altre note
Browse all plastic beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
40175 | 4061836952167 |
36742-1G | 4061831827231 |
T54402-500G | 4061838357052 |
T54402-5G | 4061838357069 |
T54402-100G | 4061838357045 |
BR87616-6EA | 4061831847734 |