Documenti fondamentali
BR81248
BRAND® burette straight stopcock
for burette capacity 25+50 mL, with spare key, PTFE key
Sinonimo/i:
Accessories, Burette stopcock, Burettes, Stopcocks
About This Item
Codice UNSPSC:
41121808
NACRES:
NB.43
Materiali
PTFE key
amber
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 81248
Descrizione generale
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
BR81248-1EA4061831861112