BR759105
BRAND® macro-cuvette
PMMA
Sinonimo/i:
Cuvettes
About This Item
Codice UNSPSC:
41121813
NACRES:
SB.32
Prodotti consigliati
Materiali
PMMA
Caratteristiche
minimum filling volume 2.5 mL
Produttore/marchio commerciale
BRAND 759105
Categorie correlate
Descrizione generale
- BRAND® Macro cuvette is manufactured under controlled room conditions.
- Ideal for the determinations in water analysis, chemistry, and life science applications.
- Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
- Clear, clean optical path with an indication of optical path orientation.
- Eliminates the risk of cross-contamination.
- Grouped by mold cavity number.
- An economical alternative to Quartz cuvettes.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)