BR759015
BRAND® semi-micro cuvette
polystyrene, minimum filling volume 1.5mL
Sinonimo/i:
Cuvettes
About This Item
Codice UNSPSC:
41121813
NACRES:
SB.32
Prodotti consigliati
Materiali
polystyrene
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
BRAND 759015
Categorie correlate
Descrizione generale
- BRAND® semi-micro cuvette is manufactured under controlled room conditions.
- Clear, clean optical path with an indication of optical path orientation.
- Packed grouped by mold cavity number to ensure the lowest variation of extinction coefficient.
- Standard 10mm light path.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)