Documenti fondamentali
BR708470
BRAND® EASYCAL micro-weighing container
Sinonimo/i:
Accessories, Calibration Software, EASYCAL
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.71
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 708470
Descrizione generale
Pipettes < 50 μL can be surprisingly easy to calibrate using EASYCAL software and the new micro-weighing container. Includes 10 filters and 3 cover caps. For use with EASYCAL Calibration software (CD-ROM) .
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
EASYCAL is a trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)