BR701362
BRAND® mat cover for deep well plate
volume 2.2 mL
Sinonimo/i:
Accessories, Cover mats, Deep-well Plates, Sample Storage
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
Prodotti consigliati
Sterilità
non-sterile
Produttore/marchio commerciale
BRAND 701362
Volume
2.2 mL
Categorie correlate
Descrizione generale
BRAND® mat cover is an ethylene vinyl acetate sealing mat for 96 deep well plates with square-shaped wells. Sealing mats are ideal for short-term storage and reliably protect samples against contamination and evaporation.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR701362-24EA | 4061831857955 |