BR65238
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 200 mm × 270 mm × 368 mm, top socket joint: 24/29
About This Item
Codice UNSPSC:
41104510
NACRES:
NB.43
Prodotti consigliati
Giunto
socket joint: top24/29
Caratteristiche
lid with socket and stopcock
Produttore/marchio commerciale
BRAND 65238
Diametro nominale × D.E. × alt.
200 mm × 270 mm × 368 mm
Categorie correlate
Descrizione generale
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid. Desiccators with sockets are supplied with 24/29 socket, and interchangeable Boro 3.3 stopcock.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR65238-1EA | 4061831857658 |