BR474702
BRAND® microscope slide
cut edges, twin frosted end
Sinonimo/i:
Microscope slides, Microscopy
About This Item
Codice UNSPSC:
41122600
NACRES:
MA.01
Prodotti consigliati
Materiali
soda-lime glass
Caratteristiche
cut edges, twin frosted end
Produttore/marchio commerciale
BRAND 474702
Misura
76 mm × 26 mm × 1 mm
Descrizione generale
- Made from clear soda-lime glass, hydrolytic class 3.
- Ground edges are recommended to reduce the risk of injury.
- Thickness approx.: 1 mm
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR474702-2500EA | 4061831857283 |